siRNA Details
Gene Symbol: Tbc1d9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134539.1 | NP_001128011.1 | 15 | 3271 | ||
XM_006255246.2 | XP_006255308.1 | 16 | 3981 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tbc1d9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134539.1 | NP_001128011.1 | 15 | 3271 | ||
XM_006255246.2 | XP_006255308.1 | 16 | 3981 |