siRNA Details
Gene Symbol: Slc45a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135868.1 | NP_001129340.1 | 5 | 1939 | ||
XM_006249765.2 | XP_006249827.1 | 5 | 1914 | |||
XM_006249766.2 | XP_006249828.1 | 5 | 2039 | |||
XM_006249767.2 | XP_006249829.1 | 5 | 2517 | |||
XM_006249769.2 | XP_006249831.1 | 5 | 2689 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145977, NM_001177628, BC024519, BC031381, AK035428, and others.
Gene Ontology Categories:
