siRNA Details
Gene Symbol: Dusp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013128.2 | NP_001013146.2 | 2 | 279 | ||
XM_006251299.2 | XP_006251361.1 | 2 | 703 | |||
GenBank mRNA | BC079285.1 | - | 2 | 261 | 7134264 |
