siRNA Details
Gene Symbol: Otx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100566.1 | NP_001094036.1 | 1047 | |||
XM_006251808.2 | XP_006251870.1 | 2358 | ||||
XM_006251809.2 | XP_006251871.1 | 2329 | ||||
XM_006251810.2 | XP_006251872.1 | 841 | ||||
XM_008770523.1 | XP_008768745.1 | 1985 | ||||
XM_008770524.1 | XP_008768746.1 | 1015 | ||||
XM_008770525.1 | XP_008768747.1 | 2173 | ||||
XM_008770526.1 | XP_008768748.1 | 1260 | ||||
XM_008770527.1 | XP_008768749.1 | 955 | ||||
XM_008770528.1 | XP_008768750.1 | 2539 | ||||
XM_008770529.1 | XP_008768751.1 | 2425 | ||||
XM_008770530.1 | XP_008768752.1 | 903 | ||||
GenBank mRNA | S81922.1 | - | 76 |
Gene Symbol: LOC100911492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751455.3 | XP_003751503.1 | 5 | 2329 | ||
XM_006251835.2 | XP_006251897.1 | 5 | 2358 | |||
XM_006251836.2 | XP_006251898.1 | 6 | 1326 | |||
XM_006251838.2 | XP_006251900.1 | 5 | 1163 | |||
XM_006251839.2 | XP_006251901.1 | 5 | 1106 | |||
XM_008770615.1 | XP_008768837.1 | 8 | 2625 | |||
XM_008770616.1 | XP_008768838.1 | 6 | 1271 | |||
XM_008770617.1 | XP_008768839.1 | 6 | 2173 | |||
XM_008770618.1 | XP_008768840.1 | 5 | 1260 | |||
XM_008770619.1 | XP_008768841.1 | 8 | 2568 | |||
XM_008770620.1 | XP_008768842.1 | 6 | 1332 | |||
XM_008770621.1 | XP_008768843.1 | 6 | 1318 | |||
XM_008770622.1 | XP_008768844.1 | 6 | 1214 | |||
XM_008770623.1 | XP_008768845.1 | 5 | 2539 | |||
XM_008770624.1 | XP_008768846.1 | 6 | 2425 |
