siRNA Details
Gene Symbol: Snx25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253168.1 | XP_006253230.1 | 15 | 2107 | ||
XM_224863.6 | XP_224863.5 | 16 | 2269 | |||
GenBank mRNA | BC086510.1 | - | 10 | 1279 | ||
FQ225871.1 | - | 4 | 403 |
