siRNA Details
Gene Symbol: Klhdc8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007685.1 | NP_001007686.1 | 4 | 964 | ||
XM_008766516.1 | XP_008764738.1 | 4 | 1358 | |||
XM_008766517.1 | XP_008764739.1 | 4 | 1051 | |||
GenBank mRNA | BC083777.1 | - | 4 | 964 | 7193594 |
