siRNA Details
Gene Symbol: Erich1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127680.1 | NP_001121152.1 | 1 | 63 | ||
XM_006253395.1 | XP_006253457.1 | 1 | 136 | |||
GenBank mRNA | BC167071.1 | - | 1 | 56 | ||
FQ227386.1 | - | 1 | 54 |
