siRNA Details
Gene Symbol: Erich1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127680.1 | NP_001121152.1 | 4 | 420 | ||
XM_006253395.1 | XP_006253457.1 | 4 | 493 | |||
GenBank mRNA | BC167071.1 | - | 4 | 413 |
