siRNA Details
Gene Symbol: Neto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107371.1 | NP_001100841.1 | 3 | 1092 | ||
XM_006255035.2 | XP_006255097.1 | 3 | 166 | |||
XM_008772133.1 | XP_008770355.1 | 3 | 166 | |||
XM_008772134.1 | XP_008770356.1 | 3 | 166 |
