siRNA Details
Gene Symbol: RGD1309362 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024884.1 | NP_001020055.1 | 2 | 1317 | ||
XM_006254759.2 | XP_006254821.1 | 2 | 1619 | |||
GenBank mRNA | BC098065.1 | - | 2 | 1317 | 7458735 |
Gene Symbol: LOC100910934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751792.3 | XP_003751840.2 | 1 | 1317 |
