siRNA Details
Gene Symbol: Arhgap26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107389.1 | NP_001100859.1 | 14 | 1574 | ||
XM_006254660.2 | XP_006254722.1 | 14 | 1643 | |||
XM_006254662.2 | XP_006254724.1 | 14 | 1639 | |||
XM_006254663.2 | XP_006254725.1 | 14 | 1638 | |||
XM_008772068.1 | XP_008770290.1 | 17 | 1872 |

More Information
Other Species Targeted

Gene Ontology Categories:
