siRNA Details
Gene Symbol: Smarca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107419.1 | NP_001100889.1 | 18 | 2189 | ||
XM_006255401.1 | XP_006255463.1 | 19 | 2980 | |||
GenBank mRNA | BC166868.1 | - | 8 | 913 |
