siRNA Details
Gene Symbol: Smarca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107419.1 | NP_001100889.1 | 22 | 2824 | ||
XM_006255401.1 | XP_006255463.1 | 23 | 3615 | |||
GenBank mRNA | BC166868.1 | - | 12 | 1548 |
