siRNA Details
Gene Symbol: Kcng4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107435.1 | NP_001100905.1 | 2 | 526 | ||
XM_006255709.2 | XP_006255771.1 | 3 | 1094 | |||
XM_006255710.2 | XP_006255772.1 | 3 | 1098 | |||
XM_006255711.2 | XP_006255773.1 | 3 | 740 | |||
XM_008772608.1 | XP_008770830.1 | 2 | 524 |
