siRNA Details
Gene Symbol: Pdcd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107452.1 | NP_001100922.1 | 3 | 315 | ||
XM_006227784.2 | XP_006227846.1 | 3 | 304 | |||
GenBank mRNA | BC168744.1 | - | 3 | 272 | 6920123 | |
FQ213638.1 | - | 3 | 236 | |||
FQ219953.1 | - | 3 | 236 | |||
FQ219997.1 | - | 3 | 236 | |||
FQ231349.1 | - | 3 | 236 | |||
FQ231760.1 | - | 3 | 236 | |||
FQ234686.1 | - | 3 | 236 |
