siRNA Details
Gene Symbol: Sphk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012066.1 | NP_001012066.1 | 6 | 1069 | ||
XM_006229085.1 | XP_006229147.1 | 6 | 1158 | |||
XM_006229086.2 | XP_006229148.1 | 6 | 1446 | |||
XM_006229088.2 | XP_006229150.1 | 6 | 1218 | |||
XM_008759383.1 | XP_008757605.1 | 6 | 1111 | |||
XM_008759384.1 | XP_008757606.1 | 6 | 1500 | |||
XM_008759385.1 | XP_008757607.1 | 6 | 1453 | |||
GenBank mRNA | BC079120.1 | - | 6 | 1069 | 7126682 | |
FQ211663.1 | - | 4 | 1598 |
