siRNA Details
Gene Symbol: Wdr73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107525.2 | NP_001100995.1 | 7 | 606 | ||
XM_006229430.2 | XP_006229492.1 | 6 | 952 | |||
GenBank mRNA | BC158658.1 | - | 7 | 606 | 8369176 |
