siRNA Details
Gene Symbol: Klhl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229818.1 | XP_006229880.1 | 3 | 5101 | ||
XM_218953.6 | XP_218953.4 | 2 | 1101 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klhl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229818.1 | XP_006229880.1 | 3 | 5101 | ||
XM_218953.6 | XP_218953.4 | 2 | 1101 |