siRNA Details
Gene Symbol: Kdm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037196.1 | NP_001032273.1 | 3, 4 | 927 | ||
XM_006230258.2 | XP_006230320.1 | 3, 4 | 1614 | |||
XM_006230259.2 | XP_006230321.1 | 3, 4 | 1791 | |||
GenBank mRNA | BC100627.1 | - | 3, 4 | 927 | 7938034 |
