siRNA Details
Gene Symbol: Tmem151a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107570.2 | NP_001101040.1 | 1 | 153 | ||
XM_003749041.3 | XP_003749089.1 | 2 | 804 | |||
GenBank mRNA | BC168914.1 | - | 1 | 153 | 7313547 |
Gene Symbol: LOC103689952 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230793.2 | XP_006230855.1 | 785 |

More Information
Other Species Targeted

