siRNA Details
Gene Symbol: Znhit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107574.1 | NP_001101044.1 | 1 | 1131 | ||
GenBank mRNA | BC168685.1 | - | 1 | 1104 | 7930608 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Znhit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107574.1 | NP_001101044.1 | 1 | 1131 | ||
GenBank mRNA | BC168685.1 | - | 1 | 1104 | 7930608 |