siRNA Details
Gene Symbol: Med23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277054.1 | NP_001263983.1 | 14 | 1575 | ||
XM_008758612.1 | XP_008756834.1 | 15 | 2366 | |||
XM_008758613.1 | XP_008756835.1 | 13 | 2251 | |||
XM_008758614.1 | XP_008756836.1 | 15 | 2367 |
