siRNA Details
Gene Symbol: Cmbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008770.3 | NP_001008770.1 | 3 | 330 | ||
XM_006232097.2 | XP_006232159.1 | 3 | 2441 | |||
GenBank mRNA | AY325197.1 | - | 3 | 332 | ||
BC088459.1 | - | 2 | 314 | 7127481 | ||
BF556107.1 | - | 3 | 337 | |||
FQ213936.1 | - | 3 | 340 | |||
FQ215832.1 | - | 2 | 332 | |||
FQ219291.1 | - | 3 | 419 | |||
FQ219760.1 | - | 3 | 343 |
