siRNA Details
Gene Symbol: Cmbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008770.3 | NP_001008770.1 | 5 | 555 | ||
XM_006232097.2 | XP_006232159.1 | 5 | 2666 | |||
GenBank mRNA | AY325197.1 | - | 5 | 557 | ||
BC088459.1 | - | 4 | 539 | 7127481 | ||
FQ209709.1 | - | 5 | 713 | |||
FQ213936.1 | - | 5 | 565 | |||
FQ215832.1 | - | 4 | 557 | |||
FQ219291.1 | - | 5 | 644 | |||
FQ219760.1 | - | 5 | 568 |
