siRNA Details
Gene Symbol: Tmem79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033896.1 | NP_001029068.1 | 2 | 514 | ||
XM_006232637.2 | XP_006232699.1 | 2 | 568 | |||
XM_008761149.1 | XP_008759371.1 | 1 | 1033 | |||
GenBank mRNA | BC101913.1 | - | 2 | 514 | 7936201 |
