siRNA Details
Gene Symbol: Vps72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107695.1 | NP_001101165.1 | 6 | 1002 | ||
XM_008761302.1 | XP_008759524.1 | 5 | 889 | |||
GenBank mRNA | BC161999.1 | - | 6 | 990 | 9036078 |
