siRNA Details
Gene Symbol: Cept1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007699.1 | NP_001007700.1 | 9 | 1434 | ||
XM_006233116.2 | XP_006233178.1 | 9 | 1362 | |||
XM_006233117.2 | XP_006233179.1 | 9 | 1362 | |||
XM_006233118.2 | XP_006233180.1 | 9 | 2199 | |||
GenBank mRNA | BC079471.1 | - | 9 | 1434 | 7122765 | |
FQ212036.1 | - | 9 | 1330 |
