siRNA Details
Gene Symbol: Med19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107741.1 | NP_001101211.1 | 2 | 437 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Med19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107741.1 | NP_001101211.1 | 2 | 437 |