siRNA Details
Gene Symbol: Med19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107741.1 | NP_001101211.1 | 3, 4 | 621 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025885, BC044302, AK010552, AK160579, AK005399, and others.
Gene Ontology Categories:
