siRNA Details
Gene Symbol: Adal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014047.1 | NP_001014069.1 | 5 | 795 | ||
XM_008762166.1 | XP_008760388.1 | 10 | 1273 | |||
XM_008762167.1 | XP_008760389.1 | 8 | 803 | |||
XM_008762168.1 | XP_008760390.1 | 6 | 704 | |||
XM_008762169.1 | XP_008760391.1 | 9 | 1009 | |||
GenBank mRNA | BC089959.1 | - | 5 | 795 | 7313149 |
