siRNA Details
Gene Symbol: Fam166a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025694.2 | NP_001020865.1 | 8, 9 | 1176 | ||
XM_008761589.1 | XP_008759811.1 | 9, 10 | 1159 | |||
XM_008761590.1 | XP_008759812.1 | 9, 10 | 1151 | |||
XM_008761591.1 | XP_008759813.1 | 8, 9 | 1099 | |||
XM_008761592.1 | XP_008759814.1 | 8, 9 | 975 | |||
XM_008761593.1 | XP_008759815.1 | 7, 8 | 1014 | |||
XM_008761594.1 | XP_008759816.1 | 6, 7 | 832 | |||
XM_008761595.1 | XP_008759817.1 | 5, 6 | 778 | |||
XM_008761596.1 | XP_008759818.1 | 9, 10 | 1229 | |||
XM_008761597.1 | XP_008759819.1 | 9, 10 | 1008 | |||
GenBank mRNA | BC097396.1 | - | 8, 9 | 1177 | 7453380 |

More Information
Other Species Targeted

