siRNA Details
Gene Symbol: Wdr91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127298.2 | NP_001120770.2 | 10 | 1445 | ||
XM_008762775.1 | XP_008760997.1 | 10 | 1547 | |||
XM_008762776.1 | XP_008760998.1 | 10 | 1846 | |||
GenBank mRNA | BC166785.1 | - | 10 | 1445 | 7935588 |
