siRNA Details
Gene Symbol: RGD1565367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270038.1 | NP_001256967.1 | 2 | 102 | ||
XM_006236264.2 | XP_006236326.1 | 2 | 1187 | |||
XM_006236267.1 | XP_006236329.1 | 1 | 61 | |||
XM_006236269.2 | XP_006236331.1 | 2 | 1027 | |||
XM_006236270.1 | XP_006236332.1 | 1 | 61 | |||
GenBank mRNA | AB511909.1 | - | 2 | 102 |
