siRNA Details
Gene Symbol: Igf2bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749741.3 | XP_003749789.1 | 12 | 1626 | ||
XM_006236474.2 | XP_006236536.1 | 12 | 1500 | |||
XM_006236475.2 | XP_006236537.1 | 10 | 1437 | |||
XM_008762933.1 | XP_008761155.1 | 12 | 1261 | |||
GenBank mRNA | AY325199.1 | - | 4 | 216 | ||
AY724479.1 | - | 12 | 1590 |
