siRNA Details
Gene Symbol: Mtmr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107880.1 | NP_001101350.1 | 7, 8 | 871 | ||
XM_006237024.2 | XP_006237086.1 | 7, 8 | 884 | |||
XM_006237025.2 | XP_006237087.1 | 6, 7 | 640 | |||
XM_006237026.1 | XP_006237088.1 | 6, 7 | 516 |
