siRNA Details
Gene Symbol: Mtmr14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107880.1 | NP_001101350.1 | 19 | 2013 | ||
XM_006237024.2 | XP_006237086.1 | 18 | 1870 | |||
XM_006237025.2 | XP_006237087.1 | 18 | 1782 | |||
XM_006237026.1 | XP_006237088.1 | 18 | 1658 |
