siRNA Details
Gene Symbol: Impad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008772.2 | NP_001008772.2 | 4, 5 | 1018 | ||
GenBank mRNA | AY321340.1 | - | 1215 |
Gene Symbol: NEWGENE_1306455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763564.1 | XP_008761786.1 | 4, 5 | 1062 |
