siRNA Details
Gene Symbol: RGD1559904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107915.1 | NP_001101385.1 | 3, 4 | 326 | ||
XM_008763542.1 | XP_008761764.1 | 3, 4 | 811 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1559904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107915.1 | NP_001101385.1 | 3, 4 | 326 | ||
XM_008763542.1 | XP_008761764.1 | 3, 4 | 811 |