siRNA Details
Gene Symbol: Abca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178095.2 | NP_835196.1 | 29 | 4318 | ||
XM_008763708.1 | XP_008761930.1 | 32 | 4907 | |||
XM_008763709.1 | XP_008761931.1 | 32 | 4907 | |||
XM_008763710.1 | XP_008761932.1 | 32 | 4900 | |||
GenBank mRNA | AY208182.1 | - | 29 | 4318 |
