siRNA Details
Gene Symbol: Rspo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107980.1 | NP_001101450.1 | 3, 4 | 631 | ||
XM_008764019.1 | XP_008762241.1 | 3, 4 | 744 | |||
XM_008764020.1 | XP_008762242.1 | 3, 4 | 797 | |||
GenBank mRNA | BC168963.1 | - | 3, 4 | 738 |
