siRNA Details
Gene Symbol: LOC103692579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764551.1 | XP_008762773.1 | 1 | 641 | ||
XM_008764552.1 | XP_008762774.1 | 1 | 641 | |||
XM_008764553.1 | XP_008762775.1 | 1 | 641 | |||
XM_008764554.1 | XP_008762776.1 | 1 | 641 |

More Information
Other Species Targeted

