siRNA Details
Gene Symbol: E2f6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100717.1 | NP_001094187.1 | 4 | 478 | ||
XM_006239913.2 | XP_006239975.1 | 5 | 525 | |||
GenBank mRNA | AY211263.1 | - | 4 | 398 | ||
AY211264.1 | - | 5 | 464 | |||
EV774540.1 | - | 1 | 47 | |||
FQ216870.1 | - | 4 | 724 |
