siRNA Details
Gene Symbol: E2f6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100717.1 | NP_001094187.1 | 3 | 262 | ||
XM_006239913.2 | XP_006239975.1 | 4 | 309 | |||
GenBank mRNA | AY211263.1 | - | 3 | 182 | ||
AY211264.1 | - | 4 | 248 | |||
FQ216870.1 | - | 3 | 508 |
