siRNA Details
Gene Symbol: Stk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108069.1 | NP_001101539.1 | 1, 2 | 1110 | ||
XM_006240910.1 | XP_006240972.1 | 1, 2 | 1105 | |||
XM_008765096.1 | XP_008763318.1 | 1, 2 | 1105 |
