siRNA Details
Gene Symbol: Csrnp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108113.1 | NP_001101583.1 | 4, 5 | 1250 | ||
XM_006242337.2 | XP_006242399.1 | 3, 4 | 793 | |||
GenBank mRNA | FQ223052.1 | - | 4, 5 | 1014 |
