siRNA Details
Gene Symbol: Gtsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079707.1 | NP_001073175.1 | 5, 6 | 533 | ||
XM_006242403.2 | XP_006242465.1 | 6, 7 | 505 | |||
XM_008765750.1 | XP_008763972.1 | 6, 7 | 495 | |||
XM_008765751.1 | XP_008763973.1 | 6, 7 | 497 | |||
XM_008765752.1 | XP_008763974.1 | 6, 7 | 566 | |||
GenBank mRNA | BC128778.1 | - | 5, 6 | 533 | 7134533 |

More Information
Other Species Targeted

