siRNA Details
Gene Symbol: Itga11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108156.1 | NP_001101626.1 | 29, 30 | 3566 | ||
XM_008766277.1 | XP_008764499.1 | 24, 25 | 3104 | |||
XM_008766278.1 | XP_008764500.1 | 29, 30 | 3941 |
