siRNA Details
Gene Symbol: Cdv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014097.1 | NP_001014119.1 | 1, 2 | 711 | ||
XM_006243659.2 | XP_006243721.1 | 1, 2 | 617 | |||
XM_006243660.2 | XP_006243722.1 | 1, 2 | 615 | |||
GenBank mRNA | BC083654.1 | - | 1, 2 | 711 | 7135192 |
