siRNA Details
Gene Symbol: Ankrd23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108211.1 | NP_001101681.1 | 2 | 298 | ||
XM_006244814.2 | XP_006244876.1 | 1 | 61 |

More Information
Other Species Targeted

Gene Ontology Categories:
