siRNA Details
Gene Symbol: Ino80d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191809.1 | NP_001178738.1 | 2 | 516 | ||
XM_006245023.2 | XP_006245085.1 | 5 | 885 | |||
XM_006245024.2 | XP_006245086.1 | 4 | 2218 | |||
XM_006245025.2 | XP_006245087.1 | 4 | 781 | |||
XM_006245026.2 | XP_006245088.1 | 5 | 1128 | |||
XM_006245029.2 | XP_006245091.1 | 5 | 995 | |||
XM_006245031.2 | XP_006245093.1 | 4 | 891 | |||
XM_006245032.2 | XP_006245094.1 | 4 | 1024 | |||
XM_006245033.2 | XP_006245095.1 | 4 | 800 | |||
XM_006245034.2 | XP_006245096.1 | 4 | 690 |
